ENA341292882 Enamine

1-[3-(ethoxycarbonyl)piperidine-1-carbonyl]-3-methyl-1H-imidazol-3-ium iodide

  • MDL number MFCD11099756
  • Cas Number : 1232233-42-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data I-
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88232

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88232
Availability details for
Quantity
  • Availability
  • Add to Cart