ENA341292885 Enamine

3-methyl-1-(2-methylpiperidine-1-carbonyl)-1H-imidazol-3-ium iodide

  • MDL number MFCD11099759
  • Cas Number : 548763-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data I-
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88235

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88235
Availability details for
Quantity
  • Availability
  • Add to Cart