ENA341292888 Enamine

3-methyl-1-(4-methylpiperidine-1-carbonyl)-1H-imidazol-3-ium iodide

  • MDL number MFCD11099762
  • Cas Number : 548763-40-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data I-
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88238

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88238
Availability details for
Quantity
  • Availability
  • Add to Cart