ENA341292889 Enamine

1-[benzyl(methyl)carbamoyl]-3-methyl-1H-imidazol-3-ium iodide

  • MDL number MFCD11099763
  • Cas Number : 380608-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data I-
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88239

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88239
Availability details for
Quantity
  • Availability
  • Add to Cart