ENA341292895 Enamine

2,2,2-trifluoroethyl N-[4-(trifluoromethoxy)phenyl]carbamate

  • MDL number MFCD11099770
  • Cas Number : 1087797-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88245

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88245
Availability details for
Quantity
  • Availability
  • Add to Cart