ENA341292905 Enamine

2,2,2-trifluoroethyl N-(2-methoxy-5-methylphenyl)carbamate

  • MDL number MFCD11099780
  • Cas Number : 1087788-53-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-88255

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88255
Availability details for
Quantity
  • Availability
  • Add to Cart