ENA341292918 Enamine

2,2,2-trifluoroethyl N-(3-chloro-2-methylphenyl)carbamate

  • MDL number MFCD11099794
  • Cas Number : 1087797-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88268

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88268
Availability details for
Quantity
  • Availability
  • Add to Cart