ENA341292920 Enamine

2,2,2-trifluoroethyl N-(3-bromophenyl)carbamate

  • MDL number MFCD01013360
  • Cas Number : 1087797-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88270

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88270
Availability details for
Quantity
  • Availability
  • Add to Cart