ENA341292934 Enamine

2,2,2-trifluoroethyl N-(5-chloropyridin-2-yl)carbamate

  • MDL number MFCD11099809
  • Cas Number : 1087788-67-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88284

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88284
Availability details for
Quantity
  • Availability
  • Add to Cart