ENA341292941 Enamine

2,2,2-trifluoroethyl N-(2-methoxy-5-nitrophenyl)carbamate

  • MDL number MFCD11099815
  • Cas Number : 1087788-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88292

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88292
Availability details for
Quantity
  • Availability
  • Add to Cart