ENA341292942 Enamine

2,2,2-trifluoroethyl N-(5,6,7,8-tetrahydronaphthalen-1-yl)carbamate

  • MDL number MFCD11099816
  • Cas Number : 1087798-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88293

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88293
Availability details for
Quantity
  • Availability
  • Add to Cart