ENA341292953 Enamine

2,2,2-trifluoroethyl N-(2,2-difluoro-1,3-dioxaindan-5-yl)carbamate

  • MDL number MFCD11099830
  • Cas Number : 1087788-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88304

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88304
Availability details for
Quantity
  • Availability
  • Add to Cart