ENA341292960 Enamine

2,2,2-trifluoroethyl N-(4-methanesulfonylphenyl)carbamate

  • MDL number MFCD11099839
  • Cas Number : 1087788-82-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88311

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88311
Availability details for
Quantity
  • Availability
  • Add to Cart