ENA341292965 Enamine

2,2,2-trifluoroethyl 2,4-dimethoxyphenylcarbamate

  • MDL number MFCD11099845
  • Cas Number : 1087788-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-88316
Availability details for
Quantity
  • Availability
  • Add to Cart