ENA341292968 Enamine

({imidazo[1,2-a]pyridin-3-yl}methyl)trimethylazanium iodide

  • MDL number MFCD09955501
  • Cas Number : 2717-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data I-
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88346

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88346
Availability details for
Quantity
  • Availability
  • Add to Cart