ENA341292970 Enamine

1-(2-fluorophenyl)-5-propyl-1H-1,2,4-triazole-3-carboxylic acid

  • MDL number MFCD12106524
  • Cas Number : 1155460-43-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137...139
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88348

Safety Information

msds https://www.enaminestore.com/catalog/EN300-88348
Availability details for
Quantity
  • Availability
  • Add to Cart