ENA341292972 Enamine

6-amino-4,4-dimethylhexanoic acid hydrochloride

  • MDL number MFCD20441600
  • Cas Number : 1354949-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88351

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88351
Availability details for
Quantity
  • Availability
  • Add to Cart