ENA341292973 Enamine

4-(2,2-difluoroethoxy)naphthalene-1-sulfonyl chloride

  • MDL number MFCD12876642
  • Cas Number : 1184189-21-6
  • Molecular Weight : 306.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-88352

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88352
Availability details for
Quantity
  • Availability
  • Add to Cart