ENA341292980 Enamine

2-propyl-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD00865742
  • Cas Number : 30709-69-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135...137
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88367

Safety Information

msds https://www.enaminestore.com/catalog/EN300-88367
Availability details for
Quantity
  • Availability
  • Add to Cart