ENA341292981 Enamine

1-(2,4-dimethyl-1,3-thiazol-5-yl)methanamine

  • MDL number MFCD09971260
  • Cas Number : 165736-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88368

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88368
Availability details for
Quantity
  • Availability
  • Add to Cart