ENA341292987 Enamine

2-(3-amino-1H-1,2,4-triazol-1-yl)-N,N-dimethylacetamide

  • MDL number MFCD12766723
  • Cas Number : 1182905-65-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88381

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88381
Availability details for
Quantity
  • Availability
  • Add to Cart