ENA341292990 Enamine

1-(2-bromoethyl)-4-nitro-1H-pyrazole hydrobromide

  • MDL number MFCD20441602
  • Cas Number : 1354954-24-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-88384

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88384
Availability details for
Quantity
  • Availability
  • Add to Cart