ENA341292992 Enamine

3-ethoxyspiro[3.5]nonan-1-amine

  • MDL number MFCD20441604
  • Cas Number : 1354952-29-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-88387

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88387
Availability details for
Quantity
  • Availability
  • Add to Cart