ENA341292997 Enamine

1-(1,3,5-trimethyl-1H-pyrazol-4-yl)propan-2-amine

  • MDL number MFCD08452614
  • Cas Number : 1007461-83-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88394

Safety Information

msds https://www.enaminestore.com/catalog/EN300-88394
Availability details for
Quantity
  • Availability
  • Add to Cart