ENA341293000 Enamine

5-tert-butyl-1-(4-chloro-2-fluorophenyl)-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD13464085
  • Cas Number : 1094411-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152...154
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88398

Safety Information

msds https://www.enaminestore.com/catalog/EN300-88398
Availability details for
Quantity
  • Availability
  • Add to Cart