ENA341293004 Enamine

[1-(6-methylpyridazin-3-yl)pyrrolidin-2-yl]methanol

  • MDL number MFCD14660979
  • Cas Number : 1251256-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88403

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88403
Availability details for
Quantity
  • Availability
  • Add to Cart