ENA341293007 Enamine

2,2,2-trifluoroethyl N-[2-(trifluoromethoxy)phenyl]carbamate

  • MDL number MFCD11099848
  • Cas Number : 1087798-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88419

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88419
Availability details for
Quantity
  • Availability
  • Add to Cart