ENA341293016 Enamine

2,2,2-trifluoroethyl N-{2-[(difluoromethyl)sulfanyl]phenyl}carbamate

  • MDL number MFCD11099856
  • Cas Number : 1087798-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88428

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88428
Availability details for
Quantity
  • Availability
  • Add to Cart