ENA341293026 Enamine

2,2,2-trifluoroethyl 4-ethoxyphenylcarbamate

  • MDL number MFCD11099866
  • Cas Number : 331-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-88438
Availability details for
Quantity
  • Availability
  • Add to Cart