ENA341293032 Enamine

2,2,2-trifluoroethyl N-(3,5-difluorophenyl)carbamate

  • MDL number MFCD11099874
  • Cas Number : 1087798-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88444

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88444
Availability details for
Quantity
  • Availability
  • Add to Cart