ENA341293041 Enamine

6-aminospiro[3.3]hept-2-ylamine dihydrochloride

  • MDL number MFCD11099887
  • Cas Number : 27259-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data 2HCl
Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-88455
Availability details for
Quantity
  • Availability
  • Add to Cart