ENA341293100 Enamine

1-[3-(trifluoromethyl)phenyl]cyclohexane-1-carboxylic acid

  • MDL number MFCD11037182
  • Cas Number : 1038719-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88538

Safety Information

msds https://www.enaminestore.com/catalog/EN300-88538
Availability details for
Quantity
  • Availability
  • Add to Cart