ENA341293121 Enamine

tert-butyl 4-(hydroxymethyl)-2,2-dimethyl-1,3-oxazolidine-3-carboxylate

  • MDL number MFCD11041411
  • Cas Number : 196964-59-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88562

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88562
Availability details for
Quantity
  • Availability
  • Add to Cart