ENA341293122 Enamine

tert-butyl 4-(iodomethyl)-2,2-dimethyl-1,3-oxazolidine-3-carboxylate

  • MDL number MFCD11099962
  • Cas Number : 1087789-13-6
  • Molecular Weight : 341.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-88563

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88563
Availability details for
Quantity
  • Availability
  • Add to Cart