ENA341293125 Enamine

tert-butyl 4-(hydroxymethyl)-2,2,5-trimethyl-1,3-oxazolidine-3-carboxylate

  • MDL number MFCD11099965
  • Cas Number : 1087798-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88566

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88566
Availability details for
Quantity
  • Availability
  • Add to Cart