ENA341293128 Enamine

1-(bromomethyl)-4-tert-butylcyclohexane

  • MDL number MFCD11847599
  • Cas Number : 92368-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88569

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88569
Availability details for
Quantity
  • Availability
  • Add to Cart