ENA341293151 Enamine

3-amino-1-ethyl-6-(trifluoromethyl)-1,2-dihydropyridin-2-one

  • MDL number MFCD11099995
  • Cas Number : 1087800-63-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88600

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88600
Availability details for
Quantity
  • Availability
  • Add to Cart