ENA341293157 Enamine

5-oxooctahydro-5H-[1,3]thiazolo[2,3-i]indole-3-carboxylic acid

  • MDL number MFCD11505637
  • Cas Number : 1189133-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88607

Safety Information

msds https://www.enaminestore.com/catalog/EN300-88607
Availability details for
Quantity
  • Availability
  • Add to Cart