ENA341293168 Enamine

3-chloro-2-methanesulfonylaniline

  • MDL number MFCD09909512
  • Cas Number : 1016863-36-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88618

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88618
Availability details for
Quantity
  • Availability
  • Add to Cart