ENA341293173 Enamine

3-ethyl-2-(morpholin-4-yl)pentanoic acid hydrochloride

  • MDL number MFCD20441609
  • Cas Number : 1354949-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88623

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88623
Availability details for
Quantity
  • Availability
  • Add to Cart