ENA341293178 Enamine

4-chlorobenzo[g]quinazoline

  • MDL number MFCD11505647
  • Cas Number : 33987-02-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178-179
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-88628

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88628
Availability details for
Quantity
  • Availability
  • Add to Cart