ENA341293183 Enamine

3-iodonaphthalene-2-carboxylic acid

  • MDL number MFCD11505652
  • Cas Number : 63212-42-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-88633

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88633
Availability details for
Quantity
  • Availability
  • Add to Cart