ENA341293191 Enamine

4-(2,3-dihydro-1,4-benzodioxin-6-yl)-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD11537281
  • Cas Number : 1096868-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88641

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88641
Availability details for
Quantity
  • Availability
  • Add to Cart