ENA341293203 Enamine

2,4-difluoro-5-(2-oxoimidazolidin-1-yl)benzoic acid

  • MDL number MFCD12525748
  • Cas Number : 1178351-23-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 246...248
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88664

Safety Information

msds https://www.enaminestore.com/catalog/EN300-88664
Availability details for
Quantity
  • Availability
  • Add to Cart