ENA341293204 Enamine

1-(2-bromoethoxy)-4-phenoxybenzene

  • MDL number MFCD00446389
  • Cas Number : 87545-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-88665

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88665
Availability details for
Quantity
  • Availability
  • Add to Cart