ENA341293207 Enamine

4-phenyl-5-(propan-2-yl)-2-(pyrrolidin-2-yl)-1H-imidazole

  • MDL number MFCD20441611
  • Cas Number : 1153105-13-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88672

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88672
Availability details for
Quantity
  • Availability
  • Add to Cart