ENA341293214 Enamine

1,4-dimethyl-1,4-diazepan-6-amine

  • MDL number MFCD19203363
  • Cas Number : 129295-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88684

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88684
Availability details for
Quantity
  • Availability
  • Add to Cart