ENA341293218 Enamine

1-(1-methyl-1H-pyrazol-4-yl)propan-1-one

  • MDL number MFCD08060047
  • Cas Number : 1007518-49-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-88721

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88721
Availability details for
Quantity
  • Availability
  • Add to Cart