ENA341293222 Enamine

methyl 3-aminopyrrolidine-1-carboxylate hydrochloride

  • MDL number MFCD20441618
  • Cas Number : 1354959-93-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160...162
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88725

Safety Information

msds https://www.enaminestore.com/catalog/EN300-88725
Availability details for
Quantity
  • Availability
  • Add to Cart