ENA341293223 Enamine

4-[2-(chloromethyl)-1,3-oxazol-5-yl]benzonitrile

  • MDL number MFCD11182417
  • Cas Number : 1094318-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124...126
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88726

Safety Information

msds https://www.enaminestore.com/catalog/EN300-88726
Availability details for
Quantity
  • Availability
  • Add to Cart